Lysing kits for Fastprep®-24 5G, Lysing Matrix E
Lieferant: MP Biomedicals
ICNA116974025EA 164 EUR
ICNA116974025 ICNA116974100 ICNA116914500 ICNA116954050 ICNA116984010 ICNA116934050 ICNA116974050 ICNA116914050 ICNA116954010 ICNA116934025 ICNA116914100
Lysing kits for Fastprep®-24 5G, Lysing Matrix E
Homogenisatoren Tischhomogenisatoren
Each impact-resistant 2 ml tube contains 1,4 mm ceramic spheres, 0,1 mm silica spheres, and one 4 mm glass bead. Lysing Matrix E tubes have purple caps and found in the FastDNA™ SPIN Kit for Soil and the FastRNA™ Pro Soil Kits. Lysing Matrix E can also be effective in lysing mixed samples, such as microbe infected tissue, and can be used in RNA extraction from tumours and other difficult tissues.