Lysing kits for Fastprep®-24 5G, Lysing Matrix SS
Lieferant: MP Biomedicals
ICNA116921100EA 319 EUR
ICNA116921100 ICNA116540431
Lysing kits for Fastprep®-24 5G, Lysing Matrix SS
Homogenisatoren Tischhomogenisatoren
Each impact-resistant 2 ml tube contains one 6,35 mm, 0,25 cal stainless-steel grinding ball.
  • Can be stored at room temperature
  • DNase and RNase free
Use to break hard/brittle samples. Specialty applications include RNA extraction from difficult samples where lysis by impaction only is preferable with minimal amounts of mechanical shearing.