Torula Yeasts
Lieferant: MP Biomedicals
ICNA0290308525EA 791 EUR
ICNA0290308525 ICNA0290308505 ICNA0290308580
Torula Yeasts
Tierbedarf Tiernahrung
Torula yeast is a common flavoring additive in processed foods such as chips, seasoning blends, and crackers.
Can be used in pet foods, in beekeeping, and as an attractant in traps to control olive flies.
Bitte beachten Store at room temperature.