Pulvertrichter
Lieferant: Hirschmann Laborgeräte
HIRS7760102EA 33 EUR
HIRS7760102 HIRS7760101 HIRS7760104 HIRS7760103 HIRS7760105
Pulvertrichter
Trichter Pulvertrichter
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable