Moisture Traps, MT Series, Agilent Technologies
Lieferant: Agilent Technologies
HEWLMT120-2EA 192 EUR
HEWLMT120-2 HEWLMT400-2 HEWLMT120-4 HEWLMT200-4 HEWLMT200-2
Moisture Traps, MT Series, Agilent Technologies
Filter Gasfilter
Indicating moisture traps are available in plastic and glass bodies.
  • Glass body traps are used when potential contaminants from plastic trap bodies are a concern
  • Glass traps are normally encased in a protective, plastic shrink wrap or a high impact plastic shield (outer trap body)
Glass and plastic bodied traps are usually pressure tested at 10342 bar, thus they are safe for use at the typical pressures required by the GC.