ADM flow meter
Lieferant: Agilent Technologies
HEWLG6691AEA 1022 EUR
HEWLG6691A
ADM flow meter
Durchflussmesser
Battery powered and portable ADM flow meter measures gas streams with changing gas composition.
  • Valuable tool for troubleshooting detector problems
  • Recalibrate annually by replacing cartridge with a NIST-certified calibration cartridge
  • Measures gas volumetrically, eliminating the need to select a gas type
  • USB connects to web interface for updates and monitoring
  • Easy to view OLED screen
  • New composite materials construction
ADM flow meters are ideal for measuring gas streams with composite gas composition. These digital flow meters are easy to maintain, by simply replacing the cartridge once a year to keep the gas flow meter compliant. There is no need to ship your meter back for recalibration, simply order and replace the cartridge yourself.
Lieferumfang: Each flow meter comes with a calibrated cartridge.