Gas line connectors for 7500
Lieferant: Agilent Technologies
HEWLG1820-65052EA 214 EUR
HEWLG1820-65052 HEWL5064-8078
Gas line connectors for 7500
Fittings und Verbinder
These gas line connectors ensure stable, consistent usage and sensitivity for a range of ICP-MS applications.
  • For plamsa and auxiliary gases
  • Carrier gas and connector for Babington nebuliser for connection with 4 mm diameter tubing
  • L-shaped PTFE connector containing a ferrule and O-ring
  • Polypropylene blend gas line connector to connect 4 mm tubing with Babington and CrossFlow nebuliser
ICP-MS systems routinely achieve parts per trillion (ppt) level sensitivity and we have a range of robust consumables and supplies, such as our precision gas line connectors for 7500 ICP-MS systems, to enhance and maintain this performance. A range of connectors are available for various nebulisers that are manufactured and tested to Agilent’s demanding specifications. Polypropylene blend gas and carrier gas line connectors are available along with associated accessories and fittings. We also provide PTFE connectors (both standard and T-shaped). Babington, CrossFlow and concentric nebulisers are covered by our high quality gas connectors. To maintain sensitivity, be sure to periodically replace consumables according to recommended maintenance schedules.