Gasreinigungsfilter
Lieferant: Agilent Technologies
VARICP7983EA 7 EUR
VARICP7983 VARICP7986SS6 VARICP7986 VARICP82117SS3 VARICP17988 VARICP17969 VARICP17984 VARICP17985 VARICP7980P6 VARICP7986SS VARICP739297 VARICP7980P4 558-0234 558-0235 558-0236 558-0237 VARICP7988P3 VARICP7987 558-0051 HEWLCP17973 VARICP7988P8 558-0053 558-0230 558-0231 558-0232 VARICP82117SS 558-0233 VARICP82117 VARICP17974 558-0048 558-0049 558-0228 569-7971 558-0229 VARICP7978 VARICP17971P
Gasreinigungsfilter
Filter Gasfilter
Clean gases reduce the risk of column damage and improve accuracy and sensitivity. These unbreakable and self-indicating filters give high sensitivity to oxygen and moisture and can be installed in seconds without tools.