Preparative HPLC cartridges
Lieferant: Agilent Technologies
HEWL895050-902EA 1683 EUR
HEWL895050-902 HEWL820212-918 HEWL870100-914 HEWL895050-909 HEWL977250-102 HEWL895050-906 HEWL877952-101 HEWL870050-902 HEWL877952-102 HEWL870150-914 HEWL977250-106 HEWL870050-906 HEWL877952-108 HEWL820212-926 HEWL770150-902 HEWL820212-925 HEWL820212-924 HEWL877150-114 HEWL820212-921 HEWL877952-105 HEWL868100-901 HEWL877952-106 HEWL820212-920 HEWL870100-906 HEWL820212-928 HEWL870100-902 HEWL897250-102 HEWL877250-112 HEWL870150-906 HEWL877250-114 HEWL870050-914 HEWL897250-109 HEWL870150-902 HEWL897250-106 HEWL897250-105 HEWL820212-933 HEWL770100-902 HEWL878150-101 HEWL820212-930 HEWL970150-902 HEWL770050-902 HEWL970150-906 HEWL895100-902 HEWL970100-906 HEWL877250-105 HEWL877250-106 HEWL877250-101 HEWL877250-102 HEWL878250-101 HEWL868050-901 HEWL895150-906 HEWL868150-901 HEWL895150-909 HEWL895150-902 HEWL877150-106 HEWL970050-902 HEWL897150-109 HEWL977150-102 HEWL970050-906 HEWL897150-106 HEWL977150-106 HEWL970100-902 HEWL895100-909 HEWL897150-102 HEWL895100-906 HEWL820212-915 HEWL820212-914 HEWL877150-102 HEWL877967-102
Preparative HPLC cartridges
Chromatographiesäulen
These cartridge columns are with reusable end fittings that have a unique design which makes them easy to install with finger-tight connections.
  • Available in a variety of bonded phases
  • Easy scale up from analytical to preparative scale without losing resolution
  • Fast preparative separation