Skimmer cones for PerkinElmer
Lieferant: Agilent Technologies
HEWL8003-0576EA 1407 EUR
HEWL8003-0576 HEWL8003-0578 HEWL8003-0572 HEWL8003-0574
Skimmer cones for PerkinElmer
Chromatographie-Zubehör Chromatographie-Ersatzteile
These skimmer and sample cones are equivalent to those from PerkinElmer ICP-MS for complete confidence.
  • Guaranteed to be compatible with PerkinElmer instruments
  • Backed by the Agilent Stand Behind Warranty that guarantees the parts will not cause instrument failure or downtime
  • Parts qualified on a genuine PerkinElmer system
Large-orifice sampler and skimmer cones provide superior long-term stability and resist clogging, allowing analysis under both high and low sample-uptake conditions. Nickel is a very rugged, long-lasting material.