Syringe filters, Econofilters
Lieferant: Agilent Technologies
HEWL5190-5310EA 1244 EUR
HEWL5190-5310 HEWL5190-5266 HEWL5190-5265 HEWL5190-5276 HEWL5190-5268 HEWL5190-5267 HEWL5190-5262 HEWL5190-5273 HEWL5190-5261 HEWL5190-5272 HEWL5190-5275 HEWL5190-5264 HEWL5190-5274 HEWL5190-5263 HEWL5190-5269 HEWL5190-5280 HEWL5190-5271 HEWL5190-5270
Syringe filters, Econofilters
Filter Spritzenvorsatzfilter
These syringe filters have a PP housing.
  • Fast and efficient filtration
  • Low level of extractables
  • High flow rate
  • Maximum sample loading capacity