Syringe filters, Captiva
Lieferant: Agilent Technologies
Captiva
HEWL5190-5134EA 221 EUR
HEWL5190-5134 HEWL5190-5133 HEWL5190-5135 HEWL5190-5132 HEWL5190-5127 HEWL5190-5126 HEWL5190-5129 HEWL5190-5128
Syringe filters, Captiva
Filter Spritzenvorsatzfilter
These syringe filters are designed for filtering samples prior to HPLC, UHPLC, and LC/MS analysis. This can improve both system performance and analytical quality.
  • Available in a wide range of sizes, formats, and membranes
Every syringe filter is manufactured using the highest-grade housing and membrane materials for excellent flow, maximum sample loading capacity, and to ensure sample integrity.