Syringe filters, Captiva, Premium
Lieferant: Agilent Technologies
Captiva
HEWL5190-5111EA 209 EUR
HEWL5190-5111 HEWL5190-5097 HEWL5190-5096 HEWL5190-5110 HEWL5190-5099 HEWL5190-5098 HEWL5190-5116 HEWL5190-5117 HEWL5190-5082 HEWL5190-5084 HEWL5190-5083 HEWL5190-5122 HEWL5190-5086 HEWL5190-5085 HEWL5190-5088 HEWL5190-5087 HEWL5190-5120 HEWL5190-5109 HEWL5190-5108 HEWL5190-5107 HEWL5190-5106 HEWL5190-5093 HEWL5190-5092 HEWL5190-5095 HEWL5190-5094 HEWL5190-5091
Syringe filters, Captiva, Premium
Filter Spritzenvorsatzfilter
These syringe filters have a PP or MBS housing.
  • Low level of extractables
  • High flow rate
  • Maximum sample loading capacity
Captiva premium syringe filters are supplied with a HPLC or LC/MS certificate.