Amino acids analysis standards, AdvanceBio AAA
Lieferant: Agilent Technologies
HEWL5061-3333EA 496 EUR
HEWL5061-3333 HEWL5062-2479 HEWL5061-3332 HEWL5190-9426 HEWL5061-3335 HEWL5061-3334 HEWL5061-3331 HEWL5061-3330 HEWL5061-3337 HEWL5062-2478 HEWL5061-3339
Amino acids analysis standards, AdvanceBio AAA
Standards Biologische Standards
A kit with amino acid standards (17 components) at different concentrations together with reagents and buffers required for analysis.