Solvent filter/degasser
Lieferant: Agilent Technologies
HEWL5188-2744EA 0 EUR
HEWL5188-2744 HEWL5188-2745 HEWL3150-0509 HEWL5188-2746 HEWL3150-0577 HEWL5188-2743 HEWL3150-0576
Solvent filter/degasser
Filter Flüssigkeits-Prozessfilter
It prevents the formation of spurious peaks within the detector due to solvent outgassing at the low-pressure end of the chromatograph.
  • Degasses eluents as particulates are removed
  • Increases solvent inlet lifetime
  • Eliminates pump downtime caused by air locks and particulates in check valves
  • Decreases piston wear, while increasing column life