In-line filters
Lieferant: Agilent Technologies
HEWL5067-1553EA 324 EUR
HEWL5067-1553 HEWLG1311-60006 HEWL5067-4638 HEWL5067-6189 HEWL280960-901 HEWL5065-4500 HEWL5064-8273 HEWL5022-2165 HEWL01090-68703 HEWL5067-1551 HEWL01090-68702
In-line filters
Filter In-Line Filter
Column inlet frit contamination that can increase column backpressure and reduce efficiency. Two types of high pressure in-line filter kits for use with any HPLC system are available.
  • To prevent blockages, always use the appropriate filters in the LC system
  • Microbore column blockages are a particular problem, due to the small diameter of the inlet frit