Sealable cuvettes
Lieferant: Hellma Analytics
634-9541EA 334 EUR
634-9541 HELL117100F-10-40 HELL117104F-10-40 HELL117200F-10-40 634-9542 HELL117200-10-40
Sealable cuvettes
Küvetten Spektralphotometer-Küvetten
Sealable cuvettes for measurements in the absence of air thanks to the airtight seal.
  • Four windows for fluorescence measurements
  • Two windows for absorption measurements
  • GL 14 screw thread cap with septum