Pulvertrichter
Lieferant: Hecht
HECH44002030EA 10.2 EUR
HECH44002030 HECH44002044
Pulvertrichter
Trichter Pulvertrichter
Powder funnels, made of PP, translucent. For decanting powdery materials. Also suitable for liquids. Cone angle 60°.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable