Digital block reactor, DRB 200
Lieferant: Hach
HACHLTV0829944001EA 2460 EUR
HACHLTV0829944001 HACHDRB200-05 HACHLTV0829942001 HACHLTV0829930001 HACHDRB200-01 HACHLTV0829940001
Digital block reactor, DRB 200
Blockthermostate Blockthermostate, digital
Digital block heaters which can hold one or two heating blocks, units allow cuvettes and reaction vessels to be digested at identical or different temperatures and time settings. Heating blocks enable cuvettes and reaction vessels to be digested at identical or different temperatures and time settings.
  • Pre-programmed for all standard digestions and freely programmable for user-specific digestions
  • Temperature stability
  • Excellent reproducibility
  • Simple handling, easy and safe to operate
  • Large, easily readable display and one-key operation for standard digestions