3-(Chlormethyl)benzoesäure ≥97%

Lieferant: Thermo Scientific
Gefahr

H60854.02EA 17.06 EUR
H60854.02 H60854.09
3-(Chlormethyl)benzoesäure ≥97%
3-(Chloromethyl)benzoic acid
Formel: C₈H₇ClO₂
Molecular Weight: 170,59
Schmelzpunkt: 136…138 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191922
CAS-Nummer: 31719-77-4
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR