Di(2-pyridyl)keton 98%
Lieferant: Thermo Scientific
|
Achtung
|
H55322.03EA
37.3
EUR
H55322.03
H55322.14
H55322.06
Di(2-pyridyl)keton 98%
Di(2-pyridyl)keton
Formel:
C₁₁H₈N₂O Molecular Weight: 184,20 g/mol Schmelzpunkt: 52…55 °C Flammpunkt: 110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006288 CAS-Nummer: 19437-26-4 EINECS: 243-067-3 |
Spezifikation Testergebnisse
Appearance (Color) | White to pale brown |
Form | Solid |
Assay (unspecified) |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...