2-(4-Methoxyphenyl)-4-thiazolcarbonsäure ≥97%
|
Achtung
|
H54606.MDEA
23.09
EUR
H54606.MD
H54606.03
H54606.06
2-(4-Methoxyphenyl)-4-thiazolcarbonsäure ≥97%
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid
Formel:
C₁₁H₉NO₃S Molecular Weight: 235,26 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03791177 CAS-Nummer: 57677-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...