2-(4-Methoxyphenyl)-4-thiazolcarbonsäure ≥97%

Achtung

H54606.MDEA 23.09 EUR
H54606.MD H54606.03 H54606.06
2-(4-Methoxyphenyl)-4-thiazolcarbonsäure ≥97%
2-(4-Methoxyphenyl)-4-thiazolecarboxylic acid
Formel: C₁₁H₉NO₃S
Molecular Weight: 235,26 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03791177
CAS-Nummer: 57677-80-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR