3-Fluor-2-methylphenylboronsäure ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
H52752.MDEA
80.3
EUR
H52752.MD
3-Fluor-2-methylphenylboronsäure ≥98%
3-Fluor-2-methylphenylboronsäure
Formel:
C₇H₈BFO₂ Molecular Weight: 153,95 g/mol Schmelzpunkt: 142…144 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368679 CAS-Nummer: 163517-61-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...