4-Hydroxy-2-(trifluormethyl)benzonitril ≥98%

Lieferant: Thermo Scientific
Achtung

H32158.03EA 212 EUR
H32158.03 H32158.MD
4-Hydroxy-2-(trifluormethyl)benzonitril ≥98%
4-Hydroxy-2-(trifluormethyl)benzonitril
Formel: C₈H₄F₃NO
Molecular Weight: 187,12 g/mol
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06797340
CAS-Nummer: 320-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR