4-Chlor-2,5-xylidin ≥98%

Lieferant: Thermo Scientific
Gefahr

H26018.06EA 119 EUR
H26018.06
4-Chlor-2,5-xylidin ≥98%
4-Chlor-2,5-xylidin
Formel: C₈H₁₀ClN
Molecular Weight: 155,63 g/mol
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08445705
CAS-Nummer: 20782-94-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR