Biobanking tubes, Cryo.s™
Lieferant: Greiner bio-one
Cryo.s™
GREI976567_960EA 1620 EUR
GREI976567_960 GREI976570_10 GREI977561_960 GREI977570_960 GREI976570_960 GREI976570MS GREI976580MS GREI976580_960 GREI976561_960 GREI976561MS 479-2163 GREI977580_960 479-2162 479-2164
Biobanking tubes, Cryo.s™
Vials
Cryo.s™ biobanking tubes are used for storage of tissue, cells, fungi, bacteria, spores, cellular extracts or body fluids at ultra-low temperature for research and development purposes in a very compact format.
  • Pre-racked
  • Non-capped tubes are non-sterile
  • Conserves upto 30% freezer space
Bitte beachten Must not be stored in the liquid phase of liquid nitrogen, but only in the gas phase. Not intended for any application in the context of reproductive medicine.