Storage Boxes, 96 Well
Lieferant: Greiner bio-one
GREI975502_1EA 34.21 EUR
GREI975502_1 GREI975502BE 391-3726 391-3727
Storage Boxes, 96 Well
Boxen Lagerboxen
The storage boxes are made of polycarbonate while the individual tubes are made of biologically inert polypropylene.
  • Comes with a coding card, which enables proper storage of samples
  • Temperature resistance of the polypropylene tubes from −80 °C to +121 °C provides for a broad range of applications