Media Bottles, tetragonal, with screw cap
Lieferant: Greiner bio-one
GREI951700_100EA 146 EUR
GREI951700_100 GREI950700_50 GREI952700_24
Media Bottles, tetragonal, with screw cap
Flaschen Zellkulturflaschen
Polyethylene terephtalate (PET), ultra-clear.
  • Thermal stability from –60 to +60°C
  • With graduation
  • Sterile
  • Free of DNase/RNase
  • Non-cytotoxic and non-pyrogenic
Media bottle with screw cap for liquid storage.