Special marker pens
Lieferant: Greiner bio-one
GREI840067_10EA 26.4 EUR
GREI841077_1 GREI840067_10
Special marker pens
Schreibstifte Labormarker
Marker pens designed for labelling all plastic articles and glassware.
  • Water-resistant
  • Quick drying
Bestellinformation: Available in four colours and two writing thicknesses.