Cell Culture Roller Bottles, CELLMASTER®
Lieferant: Greiner bio-one
CELLMASTER®
GREI680180_18EA 192 EUR
GREI680180_18 391-3170 GREI680170_30
Cell Culture Roller Bottles, CELLMASTER®
Flaschen Rollerflaschen
PET or PETG with screw caps (HDPE), sterile.
Available in short or long form and with a smooth or a ribbed surface. The ribbed design increases the growth area of the roller bottle without changing the height. Short form bottles with a ribbed design have a 2-fold or 2.5-fold expanded growth area. Long forms with a ribbed design have a 5—fold expanded growth area compared to the smooth form. All bottles are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • Surface suitable for many adherent cell lines
  • PET is very stable and gas permeable; PETG is a glycolised PET which allows more stability at a consistent level of transparency
  • Break-proof
  • All bottles have standard screw caps
Available in short or long form and with a smooth or a ribbed surface. The ribbed design increases the growth area of the roller bottle without changing the height. Short form bottles with a ribbed design have a 2-fold or 2.5-fold expanded growth area. Long forms with a ribbed design have a 5—fold expanded growth area compared to the smooth form. All bottles are certified free of RNase, DNase and human DNA, are non-pyrogenic and non-cytotoxic.
  • Surface suitable for many adherent cell lines
  • PET is very stable and gas permeable; PETG is a glycolised PET which allows more stability at a consistent level of transparency
  • Break-proof
  • All bottles have standard screw caps
Ø×H: Short form (smooth) 116×276 mm; short form (ribbed) 122×275 mm; long form 122×500 mm.
Ø×H: Short form (smooth) 116×276 mm; short form (ribbed) 122×275 mm; long form 122×500 mm
Standards: USP Class VI certified
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904