Cell culture flasks, cell-repellent surface, CELLSTAR®
Lieferant: Greiner bio-one
CELLSTAR®
GREI660985_5EA 173 EUR
GREI660985_5 GREI658980_15 GREI661980_4 GREI690985_20 GREI658985_15 GREI661985_4 GREI690980_20 GREI660980_5
Cell culture flasks, cell-repellent surface, CELLSTAR®
Kolben Zellkulturkolben
These sterile, PS cell culture flasks have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • With Standard or filter screw cap
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures and stem cell aggregate formation, suspension culture of semi-adherent and adherent cell (eg. for suspension culture of macrophages), gel-based 3D cultures, or magnetic 3D cell culturing (Nano3D technology).