Cell culture microplates, cell-repellent surface, CELLSTAR®
Lieferant: Greiner bio-one
CELLSTAR®
GREI650970_6EA 211 EUR
GREI650970_6 GREI781970_60 GREI781976_32 GREI655976_32 GREI655970_6 GREI651970_6
Cell culture microplates, cell-repellent surface, CELLSTAR®
Mikrotiterplatten Zellkultur-Platten
These sterile, PS multiwell cell culture plates have a cell-repellent surface to effectively inhibit cell attachment. Achieved through a chemical polymer modification, the surface does not degrade or leach under common cell culture conditions, rendering an ideal substrate for native cell culture experiments.
  • Effectively inhibits cell adhesion
  • Sterile
  • Lid with condensation rings
  • Other formats available (flasks, dishes, multiwell plates)
Characterised by low cell attachment, the cell-repellent surface is ideal for applications such as spheroid cultures and stem cell aggregate formation, suspension culture of semi-adherent and adherent cell (eg. for suspension culture of macrophages), gel-based 3D cultures, or magnetic 3D cell culturing (Nano3D technology).