Strips of 8 PCR tubes with attached caps
Lieferant: Greiner bio-one
GREI608281_120EA 154 EUR
GREI608281_120 GREI608286_120 GREI608283_120 GREI608284_120 GREI608281_600 GREI608201_125
Strips of 8 PCR tubes with attached caps
Röhrchen PCR-Gefäße
PP, natural.
  • Free of detectable DNase/RNase, human DNA and pyrogens
  • Free of heavy metals
  • Temperature range: –20 °C to +105 °C
Comprehensive thermal cycler compatibility.
Verpackung: Packaging labelled with product symbols for easier identification and warehousing.