Flaschen zur Wasserprobenahme, steril
Lieferant: Greiner bio-one
GREI230395EA 121 EUR
GREI250395 GREI230395 GREI250390 GREI230390_52
Flaschen zur Wasserprobenahme, steril
Flaschen Flaschen zur Wasserprobenahme
The flasks are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken.
  • Dosage in compliance with standards: ISO 5667-3/ISO 19458
  • Sodium thiosulphate in powder form
  • Tamper evident blue screw cap guarantees the sterility of the bottles prior to use