Hair and body shower gels with conditioner, Refresh™ Luxury 3 in 1 Hair & Body
Lieferant: SC Johnson
Refresh™ Luxury 3 in 1 Hair & Body
EVONCMW2LTEA 81.9 EUR
EVONCMW2LT EVONCMW1L
Hair and body shower gels with conditioner, Refresh™ Luxury 3 in 1 Hair & Body
Hautpflege Seifen und Hautreiniger
High quality, creamy, pleasant unisex fragranced hair and body shower gel removes general dirt and grime. For use at any workplace, leisure or hospitality shower facility.
  • Contains skin conditioners to improve skin hydration and prevent drying
  • Contains hair conditioner
  • Non-irritating to eyes
  • Colourant-free, perfumed
Bitte beachten *Produkte auf dieser Seite sind außerhalb des Anwendungsbereichs der EU Richtlinie 2019/904