Sample container, oblong, with lined closure
Lieferant: EP Scientific
EPSC316-0125BPCEA 2320 EUR
EPSC316-0125BPC EPSC316-0500BPC EPSC316-0250BPC EPSC316-1000BPC
Sample container, oblong, with lined closure
Probenbehälter Universalbehälter
HDPE, natural, with LDPE-lined PP cap
  • Certified to meet EPA Performance Based Specifications for metals, cyanide and fluoride analyses
  • Caps assembled onto containers
  • Space-saving shape for optimal storage efficiency
These oblong wide-mouth bottles are excellent general purpose sample containers for liquid sampling for the purpose of inorganic analyte testing. The wide mouth feature makes this bottle ideal for sampling more viscous liquid samples like industrial wastes and sludges. Space-saving oblong shape and flat side panels allow bottles to be packed tightly into coolers saving valuable space and stabilizing samples within the shipping container.
Lieferumfang: Delivery: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Lieferumfang: Supplied with Certificate of Analysis (CofA) in the case and barcoded with lot number for traceability.
Verpackung: Carton sealed with security tape providing tamper evidence.