Alcotest® 6820 handheld breathalyser specifically for the needs of law enforcement officers to keep the roads safe and free of impaired drivers.
- Fast, accurate, and reliable results
- Hygienic and easy to hold and operate with only one hand
- Resistant to rough weather conditions such as cold or moisture
- Slide'n'click" mouthpiece and easily snapped into the holder even when it’s dark
The expanded memory capacity is able to record up to 5000 test results. An optical interface enables wireless transmission of data to the mobile printer directly on site.
A LED display and acoustic signals support the visual display and report the completion of a test measurement. Even with a high alcohol content, the sensor provides fast accurate results for both active as well as passive measurements.