Rubber protection mats
Lieferant: Deutsch & Neumann
DENE5203004EA 512 EUR
DENE5203004 DENE5203015 DENE5202003 DENE5202015 DENE5202004 DENE5203002 DENE5202001 DENE5203003 DENE5202002 DENE5203001
Rubber protection mats
Oberflächenschutz Oberflächenschutz
Black protection sheets made from fluoroelastomer rubber (FPM). Durometer 75 Shore A.
  • For temperature ranges from −10 to over +200 °C
  • Good resistance to ozone and weathering
  • Benzine and mineral oil resistant