Surface protection mats
Lieferant: Deutsch & Neumann
DENE5101002EA 458 EUR
DENE5101002 DENE5101001 DENE5101004 DENE5101003 DENE5101005
Surface protection mats
Oberflächenschutz Oberflächenschutz
Translucent bench mats made from silicone rubber (VMQ).
  • Temperature range from −60 to +200° C
  • Short periods up to 260° C
  • Durometer 60 Shore A