Safety Containers
Lieferant: Deltalab
DELT232806EA 2240 EUR
DELT232806 DELT232808
Safety Containers
Behälter Abfallbehälter für spitze/scharfe Abfälle
Manufactured in PP, these containers are also shockproof, perforation and solvents resistant.
  • Shockproof, perforation and solvents resistant
  • Can be incinerated
  • Two ways of locking: Provisional and permanent
Portable models (232806) are designed to be easy opened and closed with just one hand.
Suitable for solid waste type II and III.
Type C devices
Removal of luerlock, luerslip and pen needles.
Type D devices
Five kinds of aperture: a wide one to discard elements of medium size; another one designed to remove scalpel blades; one to remove luerlock and blood collection systems; to remove butterfly and luerslip needles and the last one for insulin pen.