Disposable particle filtering respirators
Lieferant: Honeywell Safety
SuperOne
DALL1032501EA 61.3 EUR
DALL1032501 DALL1013203 DALL1013205 DALL1013204
Disposable particle filtering respirators
Atemschutzmasken Einweg-Atemschutzmasken Atemschutzmasken gegen Partikel
Ultra-light, disposable FFP half masks with elastic straps. Ideal to protect against dusts, mists and fumes.
  • Upper part pre-formed (no nose-bridge) for a quick and easy adjustment
  • Great inner volume to avoid any oppression on face and to ease communication
  • Ultra-soft inner veil to offer a soft and pleasant contact to the skin and improve the breathing comfort
  • Twin supple and resistant elastics, stapled on ear tabs to avoid any risk of irritation and of leakage in the filtering area
  • No latex, no silicone, no PVC
  • Filtration Efficiency >99,9%
Ideal for use in textile, construction, iron and steel industries.
Standards: EN 149:2001 + A1: 2009