Containers, Click Pack, nestable, with screw lid, UN-marking
Lieferant: CurTec
CURT4525-70-004EA 41.3 EUR
CURT4525-70-004 CURT4415-60-004 CURT4520-60-004_U CURT4420-60-004 CURT4515-60-004
Containers, Click Pack, nestable, with screw lid, UN-marking
Fässer, Fässer, Trommeln
Nestable containers, made of white HDPE, with dark blue screw lid. For storage and shipment of powders and pastes.
  • Water vapour tight
  • Tamper evident
  • Stackable
  • UN marking (1H2) for hazardous solids
  • Smooth inside
  • Food grade
Easy to close and open with just a quarter-turn. Take up less storage space when empty and help reduce your logistic cost.
Standards: UN-X certified (1H2) for solids