Serological pipettes, Corning®
Lieferant: Corning
Corning®
CORN7371EA 398 EUR
CORN7371 CORN7374 CORN7375
Serological pipettes, Corning®
Pipetten Serologische Pipetten
Sterile serological pipettes made of PS.
Verpackung: Triple bagged.
Bitte beachten These pipettes are "Made To Order" products. The minimum order quantity is 40 cases. Please search for Stripette® pipettes when looking for smaller order quantities.