Funnels, with short stem, Corning®
Lieferant: Corning
Corning®
CORN6120P-75EA 16.36 EUR
CORN6120P-75 CORN6120P-40 CORN6120P-120 CORN6120P-30 CORN6120P-50 CORN6120P-100 CORN6120P-150
Funnels, with short stem, Corning®
Trichter Filtertrichter Filtertrichter
Funnels, made of durable PP, translucent.
  • Very good chemical resistance
  • Autoclavable
  • Suitable for use with standard filter paper
Allow safe and easy transfers of solids or liquids from one vessel to another.