Multicoated microplates
Lieferant: Corning
Corning®
CORN3823EA 781.83 EUR
CORN3823 CORN3829
Multicoated microplates
Mikrotiterplatten Zellkultur-Platten
The Corning® multicoated microplate provides users access to six different surface treatments on a single plate.
This microplate is useful when determining the correct surface for assay requirements. Single surface microplates can then be used for the full screen or experiment. These multicoated plates are currently available in 96-well, 384-well, and 1536-well formats in black with clear bottom.