3-(N,N-Dimethylamino)phenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-8674-5GEA 1430 EUR
COBBPN-8674-5G COBBPN-8674-1G
3-(N,N-Dimethylamino)phenylboronsäurepinakolester
3-(N,N-Dimethylamino)phenylboronsäurepinakolester
Formel: C₁₄H₂₂BNO₂
Molecular Weight: 247,1400 g/mol
MDL: MFCD06795669
CAS-Nummer: 325142-87-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR