3-Methoxy-4-nitrophenylboronsäurepinakolester

Lieferant: COMBI-BLOCKS INC.

COBBPN-5785-1GEA 503 EUR
COBBPN-5785-1G COBBPN-5785-250M
3-Methoxy-4-nitrophenylboronsäurepinakolester
3-Methoxy-4-nitrophenylboronsäurepinakolester
Formel: C₁₃H₁₈BNO₅
MDL: MFCD13192152
CAS-Nummer: 755026-96-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR