3-(Methoxycarbonyl)phenylboronsäurepinakolester
Lieferant: COMBI-BLOCKS INC.
COBBPN-2116-25GEA
382
EUR
COBBPN-2116-25G
COBBPN-2116-5G
COBBPN-2116-1G
3-(Methoxycarbonyl)phenylboronsäurepinakolester
3-(Methoxycarbonyl)phenylboronsäurepinakolester
Formel:
C₁₄H₁₉BO₄ Molecular Weight: 262,1093 g/mol |
MDL:
MFCD03789258 CAS-Nummer: 480425-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...