6-Brom-3-ethyl-3H-[1,2,3]triazolo[4,5-b]pyridin

Lieferant: COMBI-BLOCKS INC.

COBBHI-1471-5GEA 571 EUR
COBBHI-1471-5G COBBHI-1471-1G
6-Brom-3-ethyl-3H-[1,2,3]triazolo[4,5-b]pyridin
6-Brom-3-ethyl-3H-[1,2,3]triazolo[4,5-b]pyridin
Formel: C₇H₇BrN₄
MDL: MFCD12913968
CAS-Nummer: 1187386-21-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR